missing translation for 'onlineSavingsMsg'
Learn More

Rutin, 97+%

Product Code. 10356970 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10356970

Brand: Thermo Scientific Acros 132391000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207671-50-9 | C27H30O16 | 610.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207671-50-9
Molecular Formula C27H30O16
Molecular Weight (g/mol) 610.52
MDL Number MFCD01319140
InChI Key IKGXIBQEEMLURG-NVPNHPEKSA-N
Synonym rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid
PubChem CID 5280805
ChEBI CHEBI:28527
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 195.0°C
Color White
Loss on Drying 5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 8438
Solubility Information Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene
Formula Weight 664.58
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Rutin hydrate, 97.0-102.0%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.