missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruv B Antibody (RuvB 4G5/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50279-0.025ml
This item is not returnable.
View return policy
Description
Ruv B Monoclonal antibody specifically detects Ruv B in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
Specifications
| Ruv B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| EC 3.1.22.4, Holliday junction branch migration complex subunit RuvB | |
| Ruv B protein from E. Coli | |
| 0.025 mL | |
| DNA Repair, Epigenetics, Virology Bacteria and Parasites | |
| E. coli | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| RuvB 4G5/1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur