missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUVBL1 Recombinant Rabbit Monoclonal Antibody (24GB1255)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557242
This item is not returnable.
View return policy
Description
RUVBL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
Specifications
| RUVBL1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RUVBL1 | |
| 2510009G06Rik; 49 kDa TATA box-binding protein-interacting protein; 49 kDa TBP-interacting protein; 54 kDa erythrocyte cytosolic protein; DNA helicase p50; ECP54; ECP-54; INO80 complex subunit H; INO80H; NMP 238; NMP238; Nuclear matrix protein 238; PONTIN; Pontin 52; Pontin52; RuvB (E coli homolog)-like 1; RuvB like AAA ATPase 1; Ruvbl1; ruvB-like 1; RuvB-like AAA ATPase 1; RuvB-like protein 1; RVB1; TAP54-alpha; TATA binding protein interacting protein 49 kDa; TIH1; Tip49; TIP49A; TIP60-a; TIP60-associated protein 54-alpha | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56505, 65137, 8607 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1255 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60122, P60123, Q9Y265 | |
| RUVBL1 | |
| A synthesized peptide derived from human TIP49A (aa 401-456). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido