missing translation for 'onlineSavingsMsg'
Learn More

RX-3117, MedChemExpress

Product Code. 16446378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446378 10 mg 10mg
16438967 5 mg 5mg
16482487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16446378 Supplier Medchem Express Supplier No. HY15228
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress RX-3117 (TV-1360) is a potent and orally active anticancer and antimetaboliteagent. RX-3117 inhibits DNA methyltransferase 1 (DNMT1). RX-3117 shows antiproliferative and anti-tumour activity. RX-3117 induces cell cycle arrest at S phase and apoptosis.

RX-3117 (TV-1360) is a potent and orally active anticancer and antimetaboliteagent. RX-3117 inhibits DNA methyltransferase 1 (DNMT1). RX-3117 shows antiproliferative and anti-tumour activity. RX-3117 induces cell cycle arrest at S phase and apoptosis.

Specifications

Chemical Name or Material RX-3117
Physical Form Solid
CAS 865838-26-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H12FN3O4
For Use With (Application) Cancer-Kinase/protease
Synonym TV-1360 fluorocyclopentenylcytosine
Solubility Information DMSO : ≥ 50 mg/mL (194.39 mM)
SMILES O=C1N([C@@H]2C(F)=C(CO)[C@@H](O)[C@H]2O)C=CC(N)=N1
Molecular Weight (g/mol) 257.22
Formula Weight 257.22
Percent Purity 98.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.