missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ryk Antibody (F35 P7 D7 F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30896-50ug
This item is not returnable.
View return policy
Description
Ryk Monoclonal antibody specifically detects Ryk in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Ryk | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| F35 P7 D7 F5 | |
| Western Blot : 1-5 μg/mL | |
| D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human Ryk protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse and rat. | |
| 50 μg | |
| Protein Kinase | |
| 6259 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction