Learn More
Invitrogen™ RYK Monoclonal Antibody (F35 P7 D7 F5)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA1615
Product Code. 11554393
  • 508.00 € / 100µg
Description
This antibody detects RYK in human, mouse, and rat samples. MA1-615 has been successfully used in Western blot, procedures. By Western blot this antibody detects the RYK receptor-like tyrosine kinase-1 in cells. The MA1-615 immunogen corresponds to the ovalbumin-conjugated synthetic peptide C (462) L T E F H A A L G A Y V(475).
RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RYK
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P34925, Q01887
RYK
Synthetic peptide corresponding to residues C(462) L T E F H A A L G A Y V(475) of RYK, conjugated to ovalbumin.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot, Immunocytochemistry
F35 P7 D7 F5
Unconjugated
RYK
AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik
Mouse
Protein G
RUO
140585, 20187, 6259
-20°C, Avoid Freeze/Thaw Cycles