missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol, 98%, Thermo Scientific Chemicals

Product Code. 15406668
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406668 5 g 5g
15496658 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406668 Supplier Thermo Scientific Alfa Aesar Supplier No. H60189.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

It is used as pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical and fine chemical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108998-83-0
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.362
MDL Number MFCD00075492
InChI Key GWVWUZJOQHWMFB-IBGZPJMESA-N
Synonym s---1,1,2-triphenylethane-1,2-diol, s---1,1,2-triphenyl-1,2-ethanediol, s-1,1,2-triphenylethane-1,2-diol, 2s-1,1,2-triphenylethane-1,2-diol, s-1,1,2-triphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2-triphenyl-, 2s, 1,1,2-triphenyl-ethane-1,2-diol, s-1,1,2-triphenyl-ethane-1,2-diol, s-triphenyl-ethane-1,2-diol, s---triphenylethylene glycol
PubChem CID 7000031
IUPAC Name (2S)-1,1,2-triphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O

Specifications

Melting Point 125°C to 127°C
Color Brown
Quantity 5 g
Beilstein 3208922
Formula Weight 290.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-1,1,2-Triphenyl-1,2-ethanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.