missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Product Code. p-7030076
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15403737

Brand: Thermo Scientific Alfa Aesar H27612.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85977-52-2 | C11H12O2 | 176.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85977-52-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368365
InChI Key VDLWTJCSPSUGOA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydro-1-naphthoic acid, s-1,2,3,4-tetrahedro-naphthoic acid, 1s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, s-1,2,3,4-tetrahydro-naphthoic acid, 1s-tetralin-1-carboxylic acid, s---1,2,3,4-tetrahydronaphthoic acid, s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, pubchem11869, s-1, 2, 3, 4-tetrahydronaphthoic acid, 1s-1,2,3,4-tetrahydro-1-naphthoic acid
PubChem CID 24820460
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 55°C to 57°C
Quantity 250 mg
Optical Rotation −12° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (S)-1,2,3,4-Tetrahydro-1-naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.