missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%

Product Code. 15408927
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15408927 250 mg 250mg
15498917 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15408927

Brand: Thermo Scientific Alfa Aesar H52295.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 93643-24-4 | C7H14N2 | 126.20 g/mol

(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93643-24-4
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD03787926
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
PubChem CID 781249
SMILES C1C[C@@H]2CNCCN2C1

Specifications

Quantity 250 mg
UN Number UN3267
Solubility Information Slightly soluble in water.
IUPAC Name (8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.