missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%

Product Code. 11389665
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389665 25 g 25g
11369665 1 g 1g
11379665 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389665 Supplier Thermo Scientific Alfa Aesar Supplier No. L19121.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66399-30-2 | C8H10FN | 139.173 g/mol

(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66399-30-2
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03092999
InChI Key QGCLEUGNYRXBMZ-LURJTMIESA-N
Synonym s-1-4-fluorophenyl ethylamine, s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s-1-4-fluoro-phenyl-ethylamine, s---1-4-fluorophenyl ethylamine, 1s-1-4-fluorophenyl ethan-1-amine, pubchem15218, 5-1-4-fluorophenyl ethanamine, s-4-fluoro-, a-methylbenzylamine
PubChem CID 2779063
IUPAC Name (1S)-1-(4-fluorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)F)N

Specifications

Melting Point -30°C
Density 1.03
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.502
Quantity 25 g
UN Number UN2735
Beilstein 6791614
Sensitivity Air Sensitive
Solubility Information Miscible with dimethyl sulfoxide.
Optical Rotation −33° (Neat)
Formula Weight 139.17
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.