missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Aminoindan, 97%

Product Code. 11965241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11965241 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11965241 Supplier Thermo Scientific Acros Supplier No. 342530010
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 61341-86-4 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61341-86-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 15.0°C
Color Yellow
Density 1.0380g/mL
Boiling Point 96.0°C (8.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H11N
Refractive Index 1.5600 to 1.5620
MDL Number MFCD00216670
Quantity 1 g
Specific Gravity 1.038
Specific Rotation + 15.62
Synonym s-+-1-aminoindan, s-1-aminoindane, s-2,3-dihydro-1h-inden-1-amine, s-+-1-aminoindane, 1s-2,3-dihydro-1h-inden-1-amine, s-+-1-indanamine, 1s-indan-1-amine, s-indan-1-ylamine, 1h-inden-1-amine, 2,3-dihydro-, 1s
Solubility Information Solubility in water: insoluble.
InChI Key XJEVHMGJSYVQBQ-UHFFFAOYNA-N
SMILES NC1CCC2=CC=CC=C12
IUPAC Name (1S)-2,3-dihydro-1H-inden-1-amine
Molecular Weight (g/mol) 133.19
PubChem CID 7000084
Formula Weight 133.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Aminoindan, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-1-Aminoindane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.