missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2,2',2' '-(10-(2-((3-((4-(Amino(carboxy)methyl)-2,6-diiodophenoxy)methyl)phenyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-2,2',2''-(10-(2-((3-((4-(Amino(carboxy)methyl)-2,6-diiodophenoxy)methyl)phenyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic Acid |
| Analyte or Component Names | (S)-2,2',2''-(10-(2-((3-((4-(Amino(carboxy)methyl)-2,6-diiodophenoxy)methyl)phenyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C31 H40 I2 N6 O10 |
| InChI Formula | InChI=1S/C31H40I2N6O10/c32-23-13-21(29(34)31(47)48)14-24(33)30(23)49-19-20-2-1-3-22(12-20)35-25(40)15-36-4-6-37(16-26(41)42)8-10-39(18-28(45)46)11-9-38(7-5-36)17-27(43)44/h1-3,12-14,29H,4-11,15-19,34H2,(H,35,40)(H,41,42)(H,43,44)(H,45,46)(H,47,48)/t29-/m0/s1 |
| Solution Type | Neat |
| SMILES | OC(CN1CCN(CC(NC2=CC=CC(COC3=C(I)C=C([C@H](N)C(O)=O)C=C3I)=C2)=O)CCN(CC(O)=O)CCN(CC(O)=O)CC1)=O |
| IUPAC Name | (S)-2,2',2''-(10-(2-((3-((4-(amino(carboxy)methyl)-2,6-diiodophenoxy)methyl)phenyl)amino)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetic acid |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 910.5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?