missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid, 98%

Product Code. 15459475
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459475 1 g 1g
15469475 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459475 Supplier Thermo Scientific Alfa Aesar Supplier No. H51964.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 122235-70-5 | C23H26N2O6 | 426.469 g/mol

It has been used in the synthesis of target chelator, the tris-catechol compound 2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the synthesis of target chelator, the tris-catechol compound 2.

Solubility
Soluble in 2ml dimethyl formamide 0.3gram.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122235-70-5
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00235897
InChI Key MVWPBNQGEGBGRF-IBGZPJMESA-N
Synonym boc-dap fmoc-oh, s-3-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino propanoic acid, s-2-boc-amino-3-fmoc-amino propionic acid, 2s-2-tert-butoxycarbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-3-9h-fluoren-9-ylmethoxy carbonyl amino, n-boc-n'-fmoc-l-2,3-diaminopropionic acid, boc-dap fmoc-oh 1g, boc-l-2,3-diaminopropionic acid fmoc, boc-dap fmoc-oh hplc, n-a-boc-n-b-fmoc-l-2,3-diamiopropionic acid
PubChem CID 7019527
IUPAC Name (2S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O

Specifications

Quantity 1 g
Beilstein 4771385
Solubility Information Soluble in 2ml dimethyl formamide 0.3gram.
Formula Weight 426.47
Percent Purity 98%
Chemical Name or Material (S)-2-(Boc-amino)-3-(Fmoc-amino)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.