missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)-4-(Fmoc-amino)butyric acid, 95%

Product Code. 15428487
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428487 5 g 5g
15459485 250 mg 250mg
15418487 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428487 Supplier Thermo Scientific Alfa Aesar Supplier No. H51989.06
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 117106-21-5 | C24H28N2O6 | 440.496 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117106-21-5
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.496
MDL Number MFCD00236845
InChI Key MJZDTTZGQUEOBL-FQEVSTJZSA-N
Synonym boc-dab fmoc-oh, s-4-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid, n-a-boc-n-g-fmoc-l-2,4-diaminobutyric acid, s-n2-boc-n4-fmoc-2,4-diaminobutyric acid, ambotzbaa1086, boc-dbu gamma-fmoc, boc-l-dab fmoc-oh, boc-l-2,4-diaminobutyric acid fmoc, n-boc-n'-fmoc-l-2,4-diaminobutyric acid, boc-dab fmoc-oh hplc
PubChem CID 2755945
IUPAC Name (2S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O

Specifications

Quantity 5 g
Beilstein 4771583
Formula Weight 440.5
Percent Purity 95%
Chemical Name or Material (S)-2-(Boc-amino)-4-(Fmoc-amino)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.