missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxy-3-phenylpropanoic acid, MedChemExpress

Product Code. 16407806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16407806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16407806 Supplier Medchem Express Supplier No. HY30220
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-2-Hydroxy-3-phenylpropanoic acid is a product of phenylalanine catabolism. An elevated level of phenyllactic acid is found in body fluids of patients with or phenylketonuria.

(S)-2-Hydroxy-3-phenylpropanoic acid is a product of phenylalanine catabolism. An elevated level of phenyllactic acid is found in body fluids of patients with or phenylketonuria.

Specifications

Chemical Name or Material (S)-2-Hydroxy-3-phenylpropanoic acid
Physical Form Solid
CAS 20312-36-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H10O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 100 mg/mL (601.79 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (200.58 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](O)CC1=CC=CC=C1
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.