missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

Product Code. 15439485
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439485 5 g 5g
15449485 250 mg 250mg
15429485 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439485 Supplier Thermo Scientific Alfa Aesar Supplier No. H51987.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 162558-25-0 | C23H26N2O6 | 426.469 g/mol

As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162558-25-0
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00235896
InChI Key PKAUMAVONPSDRW-IBGZPJMESA-N
Synonym fmoc-dap boc-oh, n-fmoc-n'-boc-l-2,3-diaminopropionic acid, fmoc-dpr boc-oh, fmoc-l-2,3-diaminopropionic acid boc, fmoc-alpha,beta-diaminopropionic acid boc-oh, 2s-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-dap boc, fmoc-dap boc, fmoc-l-dpa boc-oh, ambotzfaa1367
PubChem CID 2756103
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 5 g
Beilstein 7057792
Solubility Information Slightly soluble in water.
Formula Weight 426.47
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.