missing translation for 'onlineSavingsMsg'
Learn More

S 38093, MedChemExpress

Product Code. 16470277
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470277 50 mg 50mg
16466637 5 mg 5mg
16464946 100 mg 100mg
16463918 10 mg 10mg
16461039 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470277 Supplier Medchem Express Supplier No. HY104003
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress S 38093 is a brain-penetrant, orally active antagonist of H3 receptor, with Kis of 8.8, 1.44 and 1.2 μM for rat, mouse and human H3 receptors, respectively.

S 38093 is a brain-penetrant, orally active antagonist of H3 receptor, with Kis of 8.8, 1.44 and 1.2 μM for rat, mouse and human H3 receptors, respectively.

Specifications

Chemical Name or Material S 38093
CAS 862896-30-8
Color White
Purity Grade Notes Research
Molecular Formula C17H24N2O2
Quantity 50 mg
Solubility Information DMSO : 2 mg/mL (6.94 mM; Need ultrasonic)
SMILES O=C(N)C1=CC=C(OCCCN2CC3C(CCC3)C2)C=C1
Molecular Weight (g/mol) 288.38
Formula Weight 288.38
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.