missing translation for 'onlineSavingsMsg'
Learn More

(S)-alpha,alpha-Diphenyl-3-pyrrolidineacetamide L-Tartrate, TRC

Product Code. 30133465
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133465 10 g 10g
30007866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133465 Supplier Toronto Research Chemicals Supplier No. TRCD49188610G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134002-26-9
Molecular Formula C18 H20 N2 O . C4 H6 O6
Molecular Weight (g/mol) 430.45
Synonym 3-Pyrrolidineacetamide, α,α-diphenyl-, (3S)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), 3-Pyrrolidineacetamide, α,α-diphenyl-, (S)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), (S)-3-(1-Carbamoyl-1,1-diphenylmethyl)pyrrolidine L-tartrate, 3-(S)-(-)-(1-Carbamoyl-1,1-diphenylmethyl)pyrrolidine tartrate, 3-(S)-(1-Carbamoyl-1,1-diphenylmethyl)pyrrolidine tartrate, (S)-2,2-Diphenyl-2-(pyrrolidin-3-yl)acetamide L-tartrate
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;2,2-diphenyl-2-[(3S)-pyrrolidin-3-yl]acetamide
SMILES NC(=O)C([C@@H]1CCNC1)(c2ccccc2)c3ccccc3.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H20N2O.C4H6O6/c19-17(21)18(16-11-12-20-13-16,14-7-3-1-4-8-14)15-9-5-2-6-10-15;5-1(3(7)8)2(6)4(9)10/h1-10,16,20H,11-13H2,(H2,19,21);1-2,5-6H,(H,7,8)(H,9,10)/t16-;1-,2-/m11/s1
Formula Weight 430.174
Chemical Name or Material (S)-alpha,alpha-Diphenyl-3-pyrrolidineacetamide L-Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.