missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 98%

Product Code. 10252842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10252842 250 mg Glass Bottle 250mg
10129931 1 g Glass Bottle 1g
10289823 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10252842 Supplier Thermo Scientific Acros Supplier No. 168232500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17257-71-5 | C10H8F3O3 | 233.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17257-71-5
Molecular Formula C10H8F3O3
Molecular Weight (g/mol) 233.17
MDL Number MFCD00064200
InChI Key JJYKJUXBWFATTE-VIFPVBQESA-M
Synonym s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
PubChem CID 6992788
SMILES CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless or White
Density 1.3030g/mL
Boiling Point 116°C to 118°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula C6H5C(OCH3)(CF3)CO2H
Refractive Index 1.4720 to 1.475
Quantity 250 mg
Fieser 11,329
Merck Index 15, 6365
Specific Gravity 1.303
Specific Rotation Condition - 71.00 (20.00°C c=1.6,CH3OH)
Specific Rotation -71
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Formula Weight 234.17
Percent Purity 98%
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material (S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.