missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 99%

Product Code. 11426336
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426336 2.5 g 2.5g
11423990 500 mg 500mg
11305045 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426336 Supplier Thermo Scientific Alfa Aesar Supplier No. B23117.88
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17257-71-5 | C10H8F3O3 | 233.17 g/mol

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is used as a chiral derivatizing agent, it reacts with an alcohol or amine of unknown stereochemistry to form an ester or amide. The absolute configuration of the ester or amide is then determined by proton and/or 19F NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid is used as a chiral derivatizing agent, it reacts with an alcohol or amine of unknown stereochemistry to form an ester or amide. The absolute configuration of the ester or amide is then determined by proton and/or 19F NMR spectroscopy.

Solubility
Readily soluble in hexane, ether, THF, CH2Cl2, benzene.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17257-71-5
Molecular Formula C10H8F3O3
Molecular Weight (g/mol) 233.17
MDL Number MFCD00064200
InChI Key JJYKJUXBWFATTE-VIFPVBQESA-M
Synonym s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
PubChem CID 6992788
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
SMILES CO[C@](C([O-])=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Melting Point 43°C to 45°C
Density 1.344
Boiling Point 105°C to 107°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 2.5 g
Beilstein 3591561
Sensitivity Hygroscopic
Merck Index 14,6280
Solubility Information Readily soluble in hexane,ether,THF,CH2Cl2,benzene.
Optical Rotation −72° (c=2 in Methanol)
Formula Weight 234.18
Percent Purity 99%
Chemical Name or Material (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.