missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Alpha-Methoxy-Alpha-(trifluoromethyl)phenylacetic Acid, TRC

Product Code. 30137627
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137627 50 mg 50mg
30133117 500 mg 500mg
30005933 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137627 Supplier Toronto Research Chemicals Supplier No. TRCA57582850MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17257-71-5
Molecular Formula C10H9O3F3
Molecular Weight (g/mol) 234.17
Synonym (2S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoic Acid; (S)-(-)-3,3,3-Trifluoro-2-phenyl-2-methoxypropanoic Acid, (S)-(-)-Mosher Acid, (S)-(-)-β,β,β-Trifluoro-α-methoxyhydratropic Acid, (S)-2-Methoxy-2-phenyl-2-(trifluoromethyl)ethanoic Acid, (S)-2-Methoxy-2-trifluoromethylphenyl Acetic Acid; (S)-Methoxy(trifluoromethyl)phenylacetic Acid;
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
SMILES CO[C@](C(=O)O)(c1ccccc1)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H9F3O3/c1-16-9(8(14)15,10(11,12)13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)/t9-/m0/s1
Chemical Name or Material (S)-(-)-Alpha-Methoxy-Alpha-(trifluoromethyl)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.