missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Carbidopa, TRC

Product Code. 30367721
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367721 1 g -
30358981 100 mg 100mg
30361534 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367721 Supplier Toronto Research Chemicals Supplier No. TRCC1759851G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28860-95-9
Molecular Formula C10 H14 N2 O4
Molecular Weight (g/mol) 226.23
Synonym (αS)-α-Hydrazinyl-3,4-dihydroxy-α-methylbenzenepropanoic acid (ACI), Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, (S)- (ZCI), Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, (αS)- (9CI), Hydrocinnamic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, L- (8CI), (-)-L-α-Hydrazino-3,4-dihydroxy-α-methylhydrocinamic acid, (-)-L-α-Hydrazino-3,4-dihydroxy-α-methylhydrocinnamic acid monohydrate, (2S)-2-(Aminoazaniumyl)-3-(3,4-dihydroxyphenyl)-2-methylpropanoate, (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid, (S)-Carbidopa, 1-α-(3,4-Dihydroxybenzyl)-α-hydrazinopropionic acid, L-3-(3,4-Dihydroxyphenyl)-2-methyl-2-hydrazinopropionic acid, L-Carbidopa, L-α-(3,4-Dihydroxybenzyl)-α-hydrazinopropionic acid, L-α-Hydrazino-3,4-dihydroxy-α-methylbenzenepropanoic acid, L-α-Hydrazino-α-methyl-3,4-dihydroxyphenylpropionic acid, L-α-Hydrazino-α-methyl-β-(3,4-dihydroxyphenyl)propionic acid, L-α-Methyl-α-hydrazino-3,4-dihydroxyphenylpropionic acid, L-α-Methyl-α-hydrazino-β-(3,4-dihydroxyphenyl)propionic acid, L-α-Methyl-β-(3,4-dihydroxyphenyl)-α-hydrazinopropionic acid, L-α-Methyldopahydrazine, Cabidopa, Carbidopa, HMD, Hydrazino-α-methyldopa, Lodosin, Lodosyn, MK 486, N-Aminomethyldopa, α-Hydrazino-α-methyl-β-(3,4-dihydroxyphenyl)propionic acid, α-Methyldopahydrazine
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
SMILES C[C@@](Cc1ccc(O)c(O)c1)(NN)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)/t10-/m0/s1
Formula Weight 226.0954
Chemical Name or Material (S)-(-)-Carbidopa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.