missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ibuprofen, MedChemExpress

Product Code. 16491979
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491979 1 g 1g
16440678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491979 Supplier Medchem Express Supplier No. HY78131A
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-(+)-Ibuprofen ((S)-Ibuprofen), a S(+)-enantiomer of Ibuprofen, is a potent COX-1 and COX-2 inhibitor with IC50s of 2.1 μM and 1.6 μM, respectively. (S)-(+)-Ibuprofen has analgesic, anti-inflammatory, anticancer and antipyretic effects.

(S)-(+)-Ibuprofen ((S)-Ibuprofen), a S(+)-enantiomer of Ibuprofen, is a potent COX-1 and COX-2 inhibitor with IC50s of 2.1 μM and 1.6 μM, respectively. (S)-(+)-Ibuprofen has analgesic, anti-inflammatory, anticancer and antipyretic effects.

Specifications

Chemical Name or Material (S)-(+)-Ibuprofen
Physical Form Powder
CAS 51146-56-6
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C13H18O2
For Use With (Application) COVID-19-immunoregulation
Synonym (S)-Ibuprofen
Solubility Information DMSO : 100 mg/mL (484.78 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (484.78 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.85 mM; Need ultrasonic and warming)
SMILES OC([C@H](C1=CC=C(CC(C)C)C=C1)C)=O
Molecular Weight (g/mol) 206.28
Formula Weight 206.28
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.