missing translation for 'onlineSavingsMsg'
Learn More

(S)-Ketoprofen Acyl-(beta)-D-glucuronide, TRC

Product Code. 30010858
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010858 1 mg 1mg
30007758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010858 Supplier Toronto Research Chemicals Supplier No. TRCK2008161MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140148-26-1
Molecular Formula C22H22O9
Molecular Weight (g/mol) 430.4
Synonym 1-[(αS)-3-Benzoyl-α-methylbenzeneacetate]-β-D-glucopyranuronic αcid, (S)-Ketoprofen Glucuronide;
SMILES C[C@@H](C1=CC=CC(=C1)C(=O)C2=CC=CC=C2)C(=O)O[C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(=O)O)O)O)O

Specifications

Quantity 1 mg
Chemical Name or Material (S)-Ketoprofen Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.