missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Ketoprofen, TRC

Product Code. 30138394
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138394 500 mg 500mg
30005250 1 g 1g
30006184 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138394 Supplier Toronto Research Chemicals Supplier No. TRCK200805500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22161-81-5
Molecular Formula C16 H14 O3
Molecular Weight (g/mol) 254.28
Synonym (αS)-3-benzoyl-α-methylbenzeneacetic acid, Benzeneacetic acid, 3-benzoyl-α-methyl-, (S)-, Hydratropic acid, m-benzoyl-, (+)- (8CI), (+)-(S)-m-Benzoylhydratropic acid, (+)-2-(3-Benzoylphenyl)propionic acid, (+)-3-Benzoyl-α-methylbenzeneacetic acid, (+)-3-Benzoylhydratropic acid, (+)-Ketoprofen, (2S)-2-(3-Benzoylphenyl)propionic acid, (S)-(+)-2-(3-Benzoylphenyl)propionic acid, (S)-2-(3-Benzoylphenyl)propanoic acid, (S)-2-(3-Benzoylphenyl)propionic acid, (S)-Ketoprofen, (αS)-3-Benzoyl-α-methylbenzeneacetic acid, Arveles, Dexketoprofen, S-(+)-Ketoprofen
IUPAC Name (2S)-2-(3-benzoylphenyl)propanoic acid
SMILES C[C@H](C(=O)O)c1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/t11-/m0/s1
Formula Weight 254.0943
Chemical Name or Material (S)-(+)-Ketoprofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.