missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide (UAMC1110), TRC

Product Code. 30410674
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410674 25 mg -
30411025 2.5 mg 2.5mg
30418945 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410674 Supplier Toronto Research Chemicals Supplier No. TRCS68791025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448440-52-5
Molecular Formula C17H14F2N4O2
Molecular Weight (g/mol) 344.32
Synonym SP-13786, UAMC1110
IUPAC Name N-[2-[(2S)-2-cyano-4,4-difluoropyrrolidin-1-yl]-2-oxoethyl]quinoline-4-carboxamide
SMILES FC1(F)C[C@@H](C#N)N(C1)C(=O)CNC(=O)c2ccnc3ccccc23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H14F2N4O2/c18-17(19)7-11(8-20)23(10-17)15(24)9-22-16(25)13-5-6-21-14-4-2-1-3-12(13)14/h1-6,11H,7,9-10H2,(H,22,25)/t11-/m0/s1
Formula Weight 344.1085
Chemical Name or Material (S)-N-(2-(2-Cyano-4,4-difluoropyrrolidin-1-yl)-2-oxoethyl)quinoline-4-carboxamide (UAMC1110)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.