missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Piperidine-2-carboxylic acid, 98%

Product Code. 10102874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10102874 25 g Glass bottle 25g
10214784 1 g Glass bottle 1g
10537454 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10102874

Brand: Thermo Scientific Acros 397830250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26250-84-0 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26250-84-0
MDL Number MFCD00151904
InChI Key JQAOHGMPAAWWQO-QMMMGPOBSA-N
Synonym boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid
PubChem CID 688617
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 123.0°C to 127.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H19NO4
Quantity 25 g
Specific Rotation -62° to -65° (20°C, 589nm) (C=1, acetic acid)
Molecular Weight (g/mol) 229.27
Formula Weight 229.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-N-BOC-Piperidine-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-BOC-Piperidine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.