missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%

Product Code. 15499475
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499475 1 g 1g
15419485 250 mg 250mg
15409485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499475 Supplier Thermo Scientific Alfa Aesar Supplier No. H51975.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 136030-33-6 | C25H21NO4 | 399.446 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136030-33-6
Molecular Formula C25H21NO4
Molecular Weight (g/mol) 399.446
MDL Number MFCD00144368
InChI Key LIRBCUNCXDZOOU-QHCPKHFHSA-N
Synonym fmoc-tic-oh, fmoc-l-tic, fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-2-9h-fluoren-9-yl methoxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-+-2-9-fluorenylmethoxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, n-fmoc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-9h-fluoren-9-ylmethoxy carbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, fmoc-tic, pubchem6028
PubChem CID 978341
IUPAC Name (3S)-2-(9H-fluoren-9-ylmethoxycarbonyl)-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES C1C(N(CC2=CC=CC=C21)C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O

Specifications

Quantity 1 g
Beilstein 4827574
Optical Rotation +26.0° (c=1 in Methanol)
Formula Weight 399.45
Percent Purity 95%
Chemical Name or Material (S)-N-Fmoc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.