missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Product Code. 11905561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11905561 500 mg Glass bottle 500mg
11915561 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905561 Supplier Thermo Scientific Acros Supplier No. 441095000
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1172127-44-4 | Molecular Formula: C25H22FNO4 | Molecular Weight: 419.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1172127-44-4
Molecular Formula C25H22FNO4
Molecular Weight (g/mol) 419.45
MDL Number MFCD17019320
InChI Key VNYGHKCAXQKZEQ-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine
PubChem CID 44598473
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
SMILES CC(CC1=C(F)C=CC=C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 122.0°C to 125.0°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation − 47.00
Formula Weight 419.46
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.