missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (S)-N-FMOC-Octylglycine, 95%

Product Code. 10199121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10199121 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10199121 Supplier Thermo Scientific Chemicals Supplier No. 361490010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 193885-59-5 | C25H31NO4 | 409.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 193885-59-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 142.6°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C25H31NO4
MDL Number MFCD01311753
Quantity 1 g
Synonym fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
InChI Key LSMLSLHVRMSYPT-QHCPKHFHSA-N
SMILES CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
Molecular Weight (g/mol) 409.52
PubChem CID 23257324
Formula Weight 409.52
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-Octylglycine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.