missing translation for 'onlineSavingsMsg'
Learn More

(S)-O-Desmethyl Naproxen, TRC

Product Code. 30040898
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040898 10 mg 10mg
30008234 25 mg 25mg
18010940 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040898 Supplier Toronto Research Chemicals Supplier No. TRCD29212510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52079-10-4
Molecular Formula C13 H12 O3
Molecular Weight (g/mol) 216.23
Synonym 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (S)-, (αS)-6-Hydroxy-α-methyl-2-naphthaleneacetic acid, (+)-2-(6-Hydroxy-2-naphthyl)propionic acid, (2S)-2-(6-Hydroxynaphthalen-2-yl)propanoic acid, (S)-2-(6-Hydroxy-2-naphthyl)propionic acid, (S)-6-O-Desmethylnaproxen, 6-Demethylnaproxen, 6-Desmethylnaproxen, 6-Hydroxynaproxen, 6-O-Demethylnaproxen, O-Desmethylnaproxen, 2-Naphthaleneacetic acid, 6-hydroxy-α-methyl-, (αS)-, Naproxen Imp. A (EP), Naproxen-O-desmethyl
IUPAC Name (2S)-2-(6-hydroxynaphthalen-2-yl)propanoic acid
SMILES C[C@H](C(=O)O)c1ccc2cc(O)ccc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H12O3/c1-8(13(15)16)9-2-3-11-7-12(14)5-4-10(11)6-9/h2-8,14H,1H3,(H,15,16)/t8-/m0/s1
Formula Weight 216.0786
Percent Purity >95%
Chemical Name or Material (2S)-2-(6-Hydroxynaphthalen-2-yl)propanoic Acid (O-Demethylnaproxen)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.