missing translation for 'onlineSavingsMsg'
Learn More

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%

Product Code. 11390114
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11390114 5 g 5g
11300124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390114 Supplier Thermo Scientific Alfa Aesar Supplier No. L00577.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 22236-45-9 | C9H13NO3S2 | 247.327 g/mol

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22236-45-9
Molecular Formula C9H13NO3S2
Molecular Weight (g/mol) 247.327
MDL Number MFCD00013922
InChI Key IRNAWARRPQUZDU-UHFFFAOYSA-N
Synonym s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
PubChem CID 99226
IUPAC Name N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C

Specifications

Melting Point 167°C to 169°C
Quantity 5 g
Beilstein 2119329
Solubility Information Soluble in EtOH,THF,DMSO.
Formula Weight 247.34
Percent Purity 98%
Chemical Name or Material S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.