missing translation for 'onlineSavingsMsg'
Learn More

(S)-Willardiine, MedChemExpress

Product Code. 16461427
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461427 50 mg 50mg
16405218 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461427 Supplier Medchem Express Supplier No. HY12499

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Willardiine is a potent agonist of AMPA/kainate receptors with EC50 of 44.8 uM.

(S)-Willardiine is a potent agonist of AMPA/kainate receptors with EC50 of 44.8 uM.

Specifications

Chemical Name or Material (S)-Willardiine
Physical Form Solid
CAS 21416-43-3
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C7H9N3O4
For Use With (Application) Neuroscience-Neuromodulation
Synonym (-)-Willardiine
Solubility Information DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES N[C@@H](CN(C(N1)=O)C=CC1=O)C(O)=O
Molecular Weight (g/mol) 199.16
Formula Weight 199.16
Percent Purity 98.0%
Grade Research
Show More Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.