missing translation for 'onlineSavingsMsg'
Learn More

S0859, MedChemExpress

Product Code. 16486598
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486598 10 mg 10mg
16469167 5 mg 5mg
16412697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486598 Supplier Medchem Express Supplier No. HY15529
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).

S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).

Specifications

Chemical Name or Material S0859
CAS 1019331-10-2
Color White
Purity Grade Notes Research
Molecular Formula C29H24ClN3O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (188.67 mM)
SMILES O=C(N(CC1=CC=C(C2=CC=CC=C2S(=O)(NC#N)=O)C=C1)CC3=CC=C(C)C=C3)C4=CC=CC=C4Cl
Molecular Weight (g/mol) 530.04
Formula Weight 530.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.