missing translation for 'onlineSavingsMsg'
Learn More

S100A8 Antibody (CL11171), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07985-100ul

 View more versions of this product

Product Code. 18675038

  • 675.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

S100A8 Monoclonal antibody specifically detects S100A8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S100A8
Monoclonal
Unconjugated
PBS, pH 7.2, containing 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11171
Immunohistochemistry 1:2000 - 1:5000, Immunohistochemistry-Paraffin 1:2000 - 1:5000
60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
This antibody was developed against a recombinant protein corresponding to amino acids: LTELEKALNSIIDVYHKYSLIKGNFHAVYRDDLKKLLETECPQYIRKKGADVWFKELDINTDGAVNFQEFLILVIKM
100 μg
Cancer
6279
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.