missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A9 Antibody (CL11191), Novus Biologicals™
Mouse Monoclonal Antibody
299.00 € - 538.00 €
Specifications
| Antigen | S100A9 |
|---|---|
| Clone | CL11191 |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686686
|
Novus Biologicals
NBP3-07983-100ul |
100 μg |
538.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665818
|
Novus Biologicals
NBP3-07983-25ul |
25 μg |
299.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A9 Monoclonal antibody specifically detects S100A9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| S100A9 | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B) | |
| This antibody was developed against a recombinant protein corresponding to amino acids: CKMSQLERNIETIINTFHQYSVKLGHPDTLNQGEFKELVRKDLQNFLKKENKNEKVIEHIMEDLDTNADKQLSFEEFIMLMARLTWASHEKMHEGDEG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL11191 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.2, containing 40% glycerol | |
| 6280 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title