missing translation for 'onlineSavingsMsg'
Learn More

S100B Mouse, Clone: 4C4.9 + S100B/1012, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 478.00 €

Specifications

Antigen S100B
Clone 4C4.9 + S100B/1012
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15930848
View Documents
EnquireBio
6285-MSM3-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15940848
View Documents
EnquireBio
6285-MSM3-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

S100B
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
6285
Purified bovine brain S100 protein (4C4.9) ;Recombinant full-length human S100B protein (S100B/1012)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
10-12kDa
4C4.9 + S100B/1012
Immunohistochemistry
Unconjugated
RUO
XP_016883913
S100B
IgG2a κ
Protein A/G Purified
Cytoplasmic
Brain, Melanoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.