missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100G Antibody (S100G/7517), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23875-20ug
This item is not returnable.
View return policy
Description
S100G Monoclonal antibody specifically detects S100G in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| S100G | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| CABP, CABP1, CaBP9K, CABP9KMGC138379, CALB3calbindin D9K, calbindin 3, (vitamin D-dependent calcium-binding protein), calbindin-D9K, protein S100-G, S100 calcium binding protein G, S100 calcium-binding protein G, S100D, Vitamin D-dependent calcium-binding protein, intestinal | |
| Recombinant fragment (around aa1-79) of human S100G protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| S100G/7517 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 795 | |
| Store at 4°C. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction