Learn More
S1PR1/EDG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55133-1-AP-20UL
 View more versions of this product
Product Code. 16876595
  • 145.00 € / 20µL
Description
The antibody is specific to S1PR1.
S1P1 is a G-protein coupled receptor for the bioactive lysosphingolipid sphingosine 1-phosphate (S1P) that is coupled to the G(i) subclass of heteromeric G proteins. S1P1 signaling leads to the activation of RAC1, SRC, PTK2/FAK1 and MAP kinases. It plays an important role in cell migration via its role in the reorganization of the actin cytoskeleton and formation of lamellipodia in response to stimuli that increase the activity of the sphingosine kinase SPHK1. S1P1 is required for normal chemotaxis toward sphingosine 1-phosphate, normal embryonic heart development, and normal cardiac morphogenesis.
Specifications
Specifications
S1PR1/EDG1
Polyclonal
Unconjugated
S1PR1
CD363, CHEDG1, D1S3362, ECGF1, EDG 1, EDG1, S1P receptor 1, S1P receptor Edg 1, S1P1, S1PR1
Rabbit
Antigen Affinity Chromatography
RUO
13609, 1901
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.77 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
O08530, P21453
S1PR1
Peptide
20 μL
Primary
Mouse, Human