missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar with Chloramphenicol and Actidone

Product Code. 11993662
Click to view available options
Description:
Sabouraud Dextrose Agar with Chloramphenicol and Actidone
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11993662

Brand: Thermo Scientific™ PO0162A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Isolate dermatophytes and other fungi in clinical samples and environmental samples with this acidic selective medium.

Isolate dermatophytes, other fungi and yeasts in clinical and environmental samples with the acidic Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar with Chloramphenicol and Actidone. The low pH supports the growth of fungi while inhibiting bacterial growth in mixed samples. Fungi maintain their typical cultural appearance and thus may be readily identified according to the standard macroscopic characters described by Sabouraud1. The addition of antimicrobials to inhibit bacteria for the isolation of pathogenic fungi has been described by several workers2,3,4,5,6,7.

Use Sabouraud Dextrose Agar with Chloramphenicol and Actidone for isolation of dermatophytes and other pathogenic fungi. The medium gives reliable results with Microsporum audouini, Microsporum canis, Trichophyton mentagrophytes, Trichophyton flavum, Trichophyton rubrum and Candida albicans.

  • Suitable for identification of pathogenic fungi from material containing large numbers of other fungi or bacteria.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 10°C
Description Sabouraud Dextrose Agar with Chloramphenicol and Actidone
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.