missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar Contact Plate

Product Code. 12952598
This item is not returnable. View return policy

Product Code. 12952598

Brand: Thermo Scientific™ PO0192C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts with this formulation in a contact plate format for effortless sample surfacing.

Isolate, cultivate and differentiate dermatophytes, other fungi and yeasts using the acidic pH Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar Contact Plate. The low pH supports the growth of fungi while inhibiting bacterial growth in mixed samples. Fungi maitain their typical appearance and thus may be readily identified according to standard macroscopic characters as described by Sabouraud1. The medium gives reliable results with Microsporum audouini, Microsporum canis, Trichophytonmentagrophytes, Trichophyton flavum, Trichophyton rubrum and Candida albicans.

Features:

  • Easy-to-handle dome: shaped bottom format for effortless surface sampling.
  • Consistent performance: smooth, even agar surface ensured through specialized manufacturing process
  • Easy-to-stack providing more stability during handling and transport.
  • Ready-to-use convenience of prepared media.

Prevent agar from falling out of the dish with Thermo Scientific Contact Plates, which feature a dome-shaped bottom. The design of the contact plates makes them easy to handle and ensures the lid remains in place during handling, while being easily removed during environmental monitoring. An inner layer on the base of the plate is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits. Easy stacking provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out.

Our contact plates are manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate. Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and helps to control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi. 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, Clear
Content And Storage 2°C to 12°C
Country of Origin Wesel, Germany
Description Sabouraud Dextrose Agar Contact Plates
Format 55mm Contact Plate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 13.5g ± 5 %
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.