missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sabouraud Dextrose Agar Irradiated (Triple Wrap) Plate
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Oxoid™ Sabouraud Dextrose Agar Plate. The Sabouraud Dextrose Agar supports the growth of yeasts, fungi and molds.
Perform effortless environmental monitoring with Tryptone Soya Agar Irradiated (Triple Wrap) Plate.
- Sterile: plates are irradiated and triple wrapped in a transparent packaging.
- Consistent: automated manufacturing procedures ensure a smooth, evenly distributed layer of agar on each plate.
- Optimized: products undergo quantitative quality control testing, using a low dose inoculum to ensure optimum performance.
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Straw, clear |
| Content And Storage | 2°C to 12°C |
| Description | Sabouraud Dextrose Agar Irradiated Triple Wrap |
| Format | 55mm Plate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 13.5g ± 5 % |
| Packaging Type | Triple Wrapped - Irradiated |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction