missing translation for 'onlineSavingsMsg'
Learn More

Sacubitril hemicalcium salt, MedChemExpress

Product Code. 16421776
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16421776

Brand: Medchem Express HY15407A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sacubitril hemicalcium salt (AHU-377 hemicalcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696.

Sacubitril hemicalcium salt (AHU-377 hemicalcium salt) is a potent NEP inhibitor with an IC50 of 5 nM. Sacubitril hemicalcium salt is a component of the heart failure medicine LCZ696.

Specifications

Chemical Name or Material Sacubitril hemicalcium salt
Physical Form Solid
CAS 1369773-39-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C24H28Ca0.5NO5
For Use With (Application) Neuroscience-Neuromodulation
Synonym AHU-377 hemicalcium salt
Solubility Information DMSO : 125 mg/mL (290.35 mM; Need ultrasonic)
SMILES O=C(CCC([O-])=O)N[C@H](CC1=CC=C(C2=CC=CC=C2)C=C1)C[C@@H](C)C(OCC)=O.[0.5Ca2+]
Molecular Weight (g/mol) 430.52
Formula Weight 430.52
Percent Purity 98.41%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.