Learn More
MEDICOM Safemask™ Fit+Pro FFP3 NR Mask
Product Code. 31781384 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 240
Product Code. Size Unit Size
31781384 One Size Pack of 240
1 options
Product Code. 31781384 Supplier MEDICOM Supplier No. M63110
  • ONLINE EXCLUSIVE PRICE
    469.54 €
    392.22 € / Pack of 240
    Save 77.32 €
    16% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SafeMask Fit+Pro FFP3 mask protects against toxic aerosols and high particle concentrations while offering optimal comfort.
  • Folded 3-panel design adapts to various face shapes and sizes
  • Sealing foams on both the upper and lower panels ensure an ideal seal while providing unmatched comfort for the wearer
  • Easy to put on and adjust thanks to sliding elastic straps and a tightening ring
  • Its individual packaging helps control the risk of contamination
Specifications
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No