missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Safety Weighing Cabinet without Filtration

Product Code. 16687293 Shop All Sartorius Products
Change view
Click to view available options
Width (Metric):
500 mm
750 mm
Length (Metric):
890 mm
900 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Length (Metric) Unit Size
16687293 500 mm 890 mm Each
16363086 750 mm 900 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16687293 Supplier Sartorius Supplier No. SWC900NF
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safety Weighing Cabinets ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing. Personnel safety and reliable weighing results from a single source.

  • Appropriate protective measures must be taken to protect the health of all laboratory staff when they weigh hazardous compounds in powdered form.
  • The Sartorius SWC Safety Weighing Cabinet offers important advantages in design and construction over traditional laboratory hoods:
    • The SWC creates a contained area around the laboratory balance which prevents any air or finely powdered particulates from escaping into the operating personnel's work area.
    • At the same time, the constant inlet air velocity and the special cabinet construction both keep the air current practically turbulence-free, and, as a result, ensure consistent and repeatable weighing results.
    • The balance and weighing cabinet are perfectly matched to each other.
    • With its SWC Safety Weighing Cabinet, Sartorius has succeeded in uniting two otherwise contradictory requirements: maximum personnel protection and reliable weighing results.
    • Maximum personnel protection: Contained area around the laboratory balance which prevents any air or finely powdered particles from escaping into the operating personnel's work area
    • Reliable weighing results: Air current practically turbulence-free, and, consistent and repeatable weighing results.
    • Clear acrylic glass construction with perfect view of the balance and the sample to be weighed
    • Extremely stable base made of black granite absorbs vibrations
    • Contaminated materials can be disposed of safely in a double-bag system
    • Calibrated airflow alarm system with audit trail function sounds an alarm whenever the airflow velocity drops below a defined level
TRUSTED_SUSTAINABILITY

Specifications

Material Acrylic Glass
Length (Metric) 890 mm
Width (English) 19.7 in.
Width (Metric) 500 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.