missing translation for 'onlineSavingsMsg'
Learn More

Salermide, MedChemExpress

Product Code. 16413258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413258

Brand: Medchem Express HY101073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Salermide is an inhibitor of Sirt1 and Sirt2; can cause strong cancer-specific apoptotic cell death.

Salermide is an inhibitor of Sirt1 and Sirt2

Specifications

Chemical Name or Material Salermide
CAS 1105698-15-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H22N2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (126.75 mM)
SMILES O=C(NC1=CC=CC(/N=C/C2=C3C=CC=CC3=CC=C2O)=C1)C(C)C4=CC=CC=C4
Molecular Weight (g/mol) 394.47
Formula Weight 394.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.