missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, MedChemExpress

Product Code. 16418638
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418638 10 g 10g
16412289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16418638 Supplier Medchem Express Supplier No. HYB0811
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. Salicylamide is an analgesic and anti-pyretic agent.

Salicylamide is an inhibitor of microsomal UDP-glucuronosyltransferase. Salicylamide is an analgesic and anti-pyretic agent.

Specifications

Chemical Name or Material Salicylamide
CAS 65-45-2
Color White
Purity Grade Notes Research
Molecular Formula C7H7NO2
Quantity 10 g
Synonym 2-Hydroxybenzamide
Solubility Information DMSO : ≥ 100 mg/mL (729.18 mM) ∣H2O : 0.1 mg/mL (0.73 mM; Need ultrasonic)
SMILES O=C(N)C1=CC=CC=C1O
Molecular Weight (g/mol) 137.14
Formula Weight 137.14
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.