missing translation for 'onlineSavingsMsg'
Learn More

Salicylanilide, MedChemExpress

Product Code. 16418756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418756 Supplier Medchem Express Supplier No. HYB1408
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Salicylanilide demonstrates a wide range of biological activities including antiviral potency which can inhibit HIV virus by targeting HIV-1 integrase or reverse transcriptase.

Salicylanilide demonstrates a wide range of biological activities including antiviral potency which can inhibit HIV virus

Specifications

Chemical Name or Material Salicylanilide
Physical Form Solid
CAS 87-17-2
Color Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H11NO2
For Use With (Application) COVID-19-anti-virus
Synonym 2-Hydroxybenzanilide
Solubility Information DMSO : 170 mg/mL (797.26 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CC=CC=C1)C2=CC=CC=C2O
Molecular Weight (g/mol) 213.23
Formula Weight 213.23
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.