missing translation for 'onlineSavingsMsg'
Learn More

Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals

Product Code. 11345076 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11345076 25 g Glass bottle 25g
11335076 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345076 Supplier Thermo Scientific Acros Supplier No. 132620250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point 177.0°C to 185.0°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4C(=NOH)OH
Quantity 25 g
Beilstein 10,98
Merck Index 15,8468
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol, diethyl ether, soluble with alkalies
Formula Weight 153.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Salicylhydroxamic acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Salicylhydroxamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.