missing translation for 'onlineSavingsMsg'
Learn More

Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals

Product Code. p-191534 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11335076

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-73-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
InChI Key HBROZNQEVUILML-UHFFFAOYSA-N
Synonym salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide
PubChem CID 66644
ChEBI CHEBI:45615
IUPAC Name N,2-dihydroxybenzamide
SMILES C1=CC=C(C(=C1)C(=O)NO)O

Specifications

Melting Point 177.0°C to 185.0°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4C(=NOH)OH
Quantity 5 g
Beilstein 10,98
Merck Index 15,8468
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol, diethyl ether, soluble with alkalies
Formula Weight 153.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Salicylhydroxamic acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Salicylhydroxamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.